This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP2R5A SIRNA Assays ›
Gene Symbol:
Ppp2r5a
Gene Name:
protein phosphatase 2, regulatory subunit B', alpha
Gene Aliases:
PR61alpha
Chromosome Location:
Chr.1: 191351981 - 191397041 on Build GRCm38
UniGene ID:
Mm.205569
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s232395

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppp2r5a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144880.4 NP_659129.2 9, 10 1610
GenBank mRNA AK145397.1 - 10, 11 996
BC023062.1 - 3, 4 180
BC059026.1 - 9, 10 1610 6810754

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006243, NM_001199756, BC022474, AK312530, BC110883, and others.
This siRNA also targets the following transcripts in Rat: NM_001107891.

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of protein dephosphorylation
negative regulation of establishment of protein localization to plasma membrane
negative regulation of lipid kinase activity
protein binding
protein phosphatase type 2A regulator activity
kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header