This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ABLIM1 SIRNA Assays ›
Gene Symbol:
Ablim1
Gene Name:
actin-binding LIM protein 1
Gene Aliases:
2210411C18Rik, 2610209L21Rik, 4833406P10Rik, 9330196J19Rik, AV079770, AW060987, AW215784, Limab1, mKIAA0059
Chromosome Location:
Chr.19: 57033264 - 57239560 on Build GRCm38
UniGene ID:
Mm.217161
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s232172

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ablim1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103177.2 NP_001096647.1 14 1649
NM_001103178.2 NP_001096648.1 12 1191
NM_001290813.1 NP_001277742.1 14 1635
NM_001290815.1 NP_001277744.1 14 1649
NM_001290816.1 NP_001277745.1 13 1544
NM_178688.3 NP_848803.3 17 1887
GenBank mRNA AF404774.1 - 17 1887
AF404775.1 - 14 1409
AF404776.1 - 12 1151
AK122196.1 - 14 1649
AK143907.1 - 13 1314
AK171860.1 - 1441
BC032216.1 - 4 339
BC141083.1 - 14 1629 9055695

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: BC105867.

Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
axon guidance
lamellipodium assembly
cilium assembly
positive regulation of transcription from RNA polymerase II promoter
actin binding
protein binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0