This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM3139 SIRNA Assays ›
Gene Symbol:
Gm3139 Gm16367 Gm16513 Gm3106
Gene Name:
predicted gene 3139
predicted gene 16367
predicted gene, Gm16513
predicted gene 3106
Gene Aliases:
-
-
-
-
Chromosome Location:
Chr.5: 94478018 - 94538367 on Build GRCm38
Chromosome Location:
Chr.5|NT_187059: 468921 - 480774 on Build GRCm38
Chromosome Location:
Chr.5: 95139278 - 95195342 on Build GRCm38
Chromosome Location:
Chr.5: 94160402 - 94221093 on Build GRCm38
UniGene ID:
Mm.420028
Mm.425309
Mm.422705
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
siRNA ID s232073

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm3139
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243937.1 NP_001230866.1 3 671
NM_001243938.1 NP_001230867.1 3 631
GenBank mRNA CN721389.1 - 1 78
Gene Symbol: Gm16367
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031622.2 NP_001026793.2 3 640
GenBank mRNA AK145705.1 - 3 653
BC099967.1 - 3 640 30027969
Gene Symbol: Gm16513
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001200055.1 NP_001186984.1 651
Gene Symbol: Gm3106
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC061196.1 - 745

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0