This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF283 SIRNA Assays ›
Gene Symbol:
ZNF283
Gene Name:
zinc finger protein 283
Gene Aliases:
HZF19, HZF41
Chromosome Location:
Chr.19: 43827292 - 43853286 on Build GRCh38
UniGene ID:
Hs.652513
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s230523

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF283
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297752.1 NP_001284681.1 6 735
NM_181845.1 NP_862828.1 7 790
XM_005258784.3 XP_005258841.1 7 903
XM_005258785.3 XP_005258842.1 6 697
XM_005258786.3 XP_005258843.1 7 862
XM_017026628.1 XP_016882117.1 8 984
XM_017026629.1 XP_016882118.1 8 1025
XM_017026630.1 XP_016882119.1 8 988
XM_017026631.1 XP_016882120.1 8 855
XM_017026632.1 XP_016882121.1 8 1031
XM_017026633.1 XP_016882122.1 8 980
XM_017026634.1 XP_016882123.1 7 733
XM_017026635.1 XP_016882124.1 6 798
XM_017026636.1 XP_016882125.1 2 6503
XM_017026637.1 XP_016882126.1 6 776
XM_017026638.1 XP_016882127.1 5 570
XM_017026639.1 XP_016882128.1 7 898
GenBank mRNA AK098175.1 - 6 647
AK294852.1 - 7 813
AY166784.1 - 2 271

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0