This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CRYGB SIRNA Assays ›
Gene Symbol:
CRYGB
Gene Name:
crystallin gamma B
Gene Aliases:
CRYG2, CTRCT39
Chromosome Location:
Chr.2: 208142573 - 208146327 on Build GRCh38
UniGene ID:
Hs.248102
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s230522

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CRYGB
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005210.3 NP_005201.2 3 533
XM_017003402.1 XP_016858891.1 3 713
GenBank mRNA BC074944.2 - 3 533 30915273
BC074945.2 - 3 533 30915496
BC117384.1 - 3 533 40125935
BC117388.1 - 3 533 40125939
BY794984.2 - 3 548
KT326704.1 - 3 538

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

visual perception
lens fiber cell morphogenesis
structural constituent of eye lens

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0