This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CLEC3B SIRNA Assays ›
Gene Symbol:
CLEC3B
Gene Name:
C-type lectin domain family 3 member B
Gene Aliases:
TN, TNA
Chromosome Location:
Chr.3: 45026207 - 45036073 on Build GRCh38
UniGene ID:
Hs.476092
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s227471

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CLEC3B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308394.1 NP_001295323.1 3 597
NM_003278.2 NP_003269.2 3 694
XM_017007116.1 XP_016862605.1 2 655
XM_017007117.1 XP_016862606.1 2 1220
GenBank mRNA AK312093.1 - 3 702
BC011024.1 - 3 659 4042921
BI826174.1 - 3 596
BI826652.1 - 3 563
CR541981.1 - 3 598
CR749292.1 - 3 4576
X64559.1 - 3 691

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ossification
platelet degranulation
positive regulation of plasminogen activation
bone mineralization
cellular response to organic substance
cellular response to transforming growth factor beta stimulus
calcium ion binding
heparin binding
carbohydrate binding
kringle domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0