siRNA Details
Gene Symbol: LINC01521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120386.1 | 1 | 480 | |||
GenBank mRNA | AK000471.1 | - | 1 | 350 | ||
CR456348.1 | - | 1 | 353 |

Gene Symbol: LINC01521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120386.1 | 1 | 480 | |||
GenBank mRNA | AK000471.1 | - | 1 | 350 | ||
CR456348.1 | - | 1 | 353 |