This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BTBD3 SIRNA Assays ›
Gene Symbol:
BTBD3
Gene Name:
BTB domain containing 3
Gene Aliases:
dJ742J24.1
Chromosome Location:
Chr.20: 11890723 - 11926595 on Build GRCh38
UniGene ID:
Hs.709366
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s22633

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BTBD3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282550.1 NP_001269479.1 2, 3 312
NM_001282551.1 NP_001269480.1 2, 3 408
NM_014962.3 NP_055777.1 1, 2 730
NM_181443.3 NP_852108.2 2, 3 493
XM_017027728.1 XP_016883217.1 3, 4 641
XM_017027729.1 XP_016883218.1 3, 4 573
GenBank mRNA AB023169.1 - 1, 2 678
AK299281.1 - 2, 3 493
BC093677.1 - 1, 2 201
BC101527.1 - 1, 2 200 8069033
BC109315.1 - 2, 3 341 40036410
BC109316.1 - 2, 3 341 40036411
CB961606.1 - 2, 3 414
DC314595.1 - 2, 3 408
DC418438.1 - 2, 3 312

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cerebral cortex development
regulation of proteolysis
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
dendrite morphogenesis
ubiquitin protein ligase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0