This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C8ORF31 SIRNA Assays ›
Gene Symbol:
C8orf31
Gene Name:
chromosome 8 open reading frame 31
Gene Aliases:
-
Chromosome Location:
Chr.8: 143038983 - 143054303 on Build GRCh38
UniGene ID:
Hs.660382
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s226226

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C8orf31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173687.3 NP_775958.1 6 1263
NR_126398.1 8 1445
NR_126399.1 6 1254
GenBank mRNA AK094450.1 - 6 1211
BC120993.2 - 6 1189

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header