This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF13 SIRNA Assays ›
Gene Symbol:
RNF13
Gene Name:
ring finger protein 13
Gene Aliases:
RZF
Chromosome Location:
Chr.3: 149812688 - 149962139 on Build GRCh38
UniGene ID:
Hs.12333
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s225498

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007282.4 NP_009213.1 8 1299
NM_183381.2 NP_899237.1 7 1197
XM_005247092.3 XP_005247149.1 7 1156
XM_011512373.2 XP_011510675.1 9 1304
XM_011512374.2 XP_011510676.1 8 1202
XM_011512376.2 XP_011510678.1 7 607
XM_017005654.1 XP_016861143.1 8 1378
XM_017005655.1 XP_016861144.1 7 1279
XM_017005656.1 XP_016861145.1 8 1258
XM_017005657.1 XP_016861146.1 8 1391
XM_017005658.1 XP_016861147.1 7 1289
XM_017005659.1 XP_016861148.1 6 1026
XM_017005660.1 XP_016861149.1 6 1016
GenBank mRNA AF037204.1 - 7 666
AF070558.1 - 7 657
AK090638.1 - 7 1177
AK090771.1 - 4 286
AK313304.1 - 7 681
BC009781.1 - 7 674 3683407
BC009803.1 - 8 777 4274992
BC017878.1 - 7 639
BG546092.1 - 5 470
BM556246.1 - 6 552
CB109037.1 - 1 24
CR456804.1 - 6 515

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein autoubiquitination
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0