This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01587 SIRNA Assays ›
Gene Symbol:
LINC01587
Gene Name:
long intergenic non-protein coding RNA 1587
Gene Aliases:
C4orf6, aC1
Chromosome Location:
Chr.4: 5524569 - 5527801 on Build GRCh38
UniGene ID:
Hs.177972
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s225422

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01587
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_126518.1 1, 2 339
NR_126519.1 2, 3 248
GenBank mRNA BC117444.1 - 1, 2 293 40125995
BC117446.1 - 1, 2 293 40125997
BX094519.1 - 1, 2 157
D82070.1 - 1, 2 339
DB043858.1 - 2, 3 248

Back To Top

More Information


Gene Ontology Categories:

Function(s)

nervous system development

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0