This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPR52 SIRNA Assays ›
Gene Symbol:
GPR52
Gene Name:
G protein-coupled receptor 52
Gene Aliases:
-
Chromosome Location:
Chr.1: 174448074 - 174449545 on Build GRCh38
UniGene ID:
Hs.673850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s225108

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GPR52
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005684.4 NP_005675.3 1 1031
GenBank mRNA BC067456.1 - 1 993 6971753
BC067457.1 - 1 993 6971754
BC067458.1 - 1 993 6971755
BC067459.1 - 1 993 6971756
BC067460.1 - 1 993 6971757
BC069460.1 - 1 993 7262282
BC095532.1 - 1 1032 7002024
DA174341.1 - 1 219
DB168074.1 - 1 65
HQ995526.1 - 1 993

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

synaptic transmission, dopaminergic
G-protein coupled receptor signaling pathway
adenylate cyclase-activating dopamine receptor signaling pathway
learning
phospholipase C-activating dopamine receptor signaling pathway
dopamine neurotransmitter receptor activity, coupled via Gs
G-protein coupled receptor activity
dopamine binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0