This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RELT SIRNA Assays ›
Gene Symbol:
RELT
Gene Name:
RELT tumor necrosis factor receptor
Gene Aliases:
TNFRSF19L, TRLT
Chromosome Location:
Chr.11: 73376360 - 73397474 on Build GRCh38
UniGene ID:
Hs.533720
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s224978

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RELT
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032871.3 NP_116260.2 11 2440
NM_152222.1 NP_689408.1 11 2378
XM_005274356.2 XP_005274413.1 6 2541
XM_006718714.2 XP_006718777.1 9 2005
XM_011545305.1 XP_011543607.1 10 2263
GenBank mRNA AK027899.1 - 11 2378
AK057036.1 - 5 2182
AK074128.1 - 3 4113
BC017279.2 - 11 2303 4873951
BC051810.1 - 11 2329 5216988

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
immune response
multicellular organism development
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
regulation of apoptotic process
positive regulation of MAPK cascade
apoptotic signaling pathway
tumor necrosis factor-activated receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0