This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTGER4 SIRNA Assays ›
Gene Symbol:
Ptger4
Gene Name:
prostaglandin E receptor 4 (subtype EP4)
Gene Aliases:
EP4, Ptger, Ptgerep4
Chromosome Location:
Chr.2: 54951630 - 54966450 on Build Rnor_6.0
UniGene ID:
Rn.16062
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s222878

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptger4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032076.3 NP_114465.3 1 158
XM_006231996.2 XP_006232058.1 2 2727
GenBank mRNA D28860.1 - 2 338
U94709.1 - 1 158

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune response
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
JNK cascade
response to mechanical stimulus
regulation of ossification
response to lipopolysaccharide
negative regulation of integrin activation
T-helper cell differentiation
negative regulation of cytokine secretion
positive regulation of cytokine secretion
negative regulation of inflammatory response
positive regulation of inflammatory response
regulation of stress fiber assembly
bone development
ERK1 and ERK2 cascade
cellular response to mechanical stimulus
negative regulation of eosinophil extravasation
prostaglandin E receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0