This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAMSTR SIRNA Assays ›
Gene Symbol:
Mamstr
Gene Name:
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.1: 101621704 - 101628379 on Build Rnor_6.0
UniGene ID:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s222772

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mamstr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001076631.5 XP_001076631.2 5 1316
XM_006229210.2 XP_006229272.1 5 1701
XM_006229211.2 XP_006229273.1 5 1260

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of myotube differentiation
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, RNA polymerase II transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header