This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ATG10 SIRNA Assays ›
Gene Symbol:
Atg10
Gene Name:
autophagy related 10
Gene Aliases:
-
Chromosome Location:
Chr.2: 19853546 - 20152900 on Build Rnor_6.0
UniGene ID:
Rn.21798
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s222648

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Atg10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109505.1 NP_001102975.1 3 201
XM_006231756.2 XP_006231818.1 4 468
XM_006231757.2 XP_006231819.1 3 1239
XM_006231758.2 XP_006231820.1 3 217
XM_006231759.2 XP_006231821.1 3 237

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein lipidation
autophagy
biological_process
positive regulation of protein modification process
protein modification by small protein conjugation
autophagy in response to ER overload
molecular_function
Atg12 transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0