This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDUFB4 SIRNA Assays ›
Gene Symbol:
Ndufb4
Gene Name:
NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4
Gene Aliases:
-
Chromosome Location:
Chr.11: 65960277 - 65966899 on Build Rnor_6.0
UniGene ID:
Rn.98986
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s222269

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndufb4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037338.1 NP_001032415.1 1 144
GenBank mRNA CB732095.1 - 1 144
FQ217067.1 - 1 151

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026610, AK004958, BC056229, BC096421, BC087907, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header