This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other XIAP SIRNA Assays ›
Gene Symbol:
Xiap
Gene Name:
X-linked inhibitor of apoptosis
Gene Aliases:
Api3, Birc4, riap3
Chromosome Location:
Chr.X: 128409424 - 128456025 on Build Rnor_6.0
UniGene ID:
Rn.91239
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s222076

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Xiap
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022231.2 NP_071567.1 3 1079
XM_006257506.2 XP_006257568.1 2 905
XM_006257508.2 XP_006257570.1 2 1019
XM_006257509.2 XP_006257571.1 2 920
XM_008773529.1 XP_008771751.1 3 1046
GenBank mRNA AB033366.1 - 3 1078
AF183429.1 - 1 749
AF304334.1 - 1674

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001204401, NM_001167, BC032729, AK313883, U32974.
This siRNA also targets the following transcripts in Mouse: NM_009688, U36842, AK147447, BC145861, BC138528, and others.

Gene Ontology Categories:

Function(s) Process(es)

cellular response to DNA damage stimulus
Wnt signaling pathway
protein ubiquitination
positive regulation of protein ubiquitination
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of neuron apoptotic process
neuron apoptotic process
positive regulation of canonical Wnt signaling pathway
mitotic spindle assembly
positive regulation of protein linear polyubiquitination
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
protease binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0