This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FXYD1 SIRNA Assays ›
Gene Symbol:
Fxyd1
Gene Name:
FXYD domain-containing ion transport regulator 1
Gene Aliases:
Plm
Chromosome Location:
Chr.1: 89484197 - 89488278 on Build Rnor_6.0
UniGene ID:
Rn.3828
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s222050

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fxyd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031648.2 NP_113836.2 5, 6 289
XM_006228838.2 XP_006228900.1 6, 7 509
XM_006228839.2 XP_006228901.1 5, 6 284
XM_008759184.1 XP_008757406.1 5, 6 298
GenBank mRNA CB712570.1 - 5, 6 289
FQ211349.1 - 5, 6 300
FQ224131.1 - 5, 6 252
FQ224338.1 - 5, 6 335
U72246.1 - 4, 5 203

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_052992, NM_019503, NM_052991, NM_194321, CA459152, and others.

Gene Ontology Categories:

Function(s) Process(es)

brain development
positive regulation of organic acid transport
regulation of cytosolic calcium ion concentration
regulation of cardiac muscle cell contraction
regulation of cardiac muscle cell membrane potential
chloride transmembrane transport
positive regulation of sodium ion export from cell
positive regulation of inorganic anion transmembrane transport
regulation of sodium ion transmembrane transporter activity
chloride channel activity
protein binding
myosin binding
sodium channel regulator activity
ATPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0