This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VOM1R110 SIRNA Assays ›
Gene Symbol:
Vom1r110 LOC100911644 Vom1r27
Gene Name:
vomeronasal 1 receptor 110
vomeronasal type-1 receptor 4-like
vomeronasal 1 receptor 27
Gene Aliases:
V1re18
-
V1re19
Chromosome Location:
Chr.1: 63184734 - 63185651 on Build Rnor_6.0
Chromosome Location:
Chr.1: 68649663 - 68650566 on Build Rnor_6.0
Chromosome Location:
Chr.1: 68528976 - 68529893 on Build Rnor_6.0
UniGene ID:
Rn.129439
-
Rn.129439
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s221733

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vom1r110
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008953.1 NP_001008953.1 1 93
GenBank mRNA AY510352.1 - 1 93
AY510356.1 - 93
Gene Symbol: LOC100911644
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008758880.1 XP_008757102.1 1 93
Gene Symbol: Vom1r27
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008952.1 NP_001008952.1 1 93
GenBank mRNA AY510351.1 - 1 93

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
pheromone receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0