This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VOM1R95 SIRNA Assays ›
Gene Symbol:
Vom1r95 Vom1r98
Gene Name:
vomeronasal 1 receptor 95
vomeronasal 1 receptor 98
Gene Aliases:
V1ra7, Vmn1r52
V1ra8
Chromosome Location:
Chr.4: 121821837 - 121822781 on Build Rnor_6.0
Chromosome Location:
Chr.4: 121912352 - 121913296 on Build Rnor_6.0
UniGene ID:
Rn.129646
Rn.129644
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s221730

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vom1r95
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008950.1 NP_001008950.1 1 509
GenBank mRNA AY510341.1 - 1 509
Gene Symbol: Vom1r98
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008948.1 NP_001008948.1 1 509
GenBank mRNA AY510365.1 - 1 509

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
response to pheromone
pheromone receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0