This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARIH2 SIRNA Assays ›
Gene Symbol:
Arih2
Gene Name:
ariadne RBR E3 ubiquitin protein ligase 2
Gene Aliases:
ARI2, TRIAD1
Chromosome Location:
Chr.8: 117387004 - 117446175 on Build Rnor_6.0
UniGene ID:
Rn.9570
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s221258

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arih2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012275.2 NP_001012275.1 6 700
XM_006243784.2 XP_006243846.1 6 1168
XM_006243785.2 XP_006243847.1 6 1165
XM_006243787.2 XP_006243849.1 5 389
XM_008766537.1 XP_008764759.1 6 853
XM_008766538.1 XP_008764760.1 6 856
GenBank mRNA CV126331.1 - 2 204

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
ubiquitin-dependent protein catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
developmental cell growth
protein K63-linked ubiquitination
protein K48-linked ubiquitination
hematopoietic stem cell proliferation
nucleic acid binding
ubiquitin-protein transferase activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0