This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC39A14 SIRNA Assays ›
Gene Symbol:
Slc39a14
Gene Name:
solute carrier family 39 (zinc transporter), member 14
Gene Aliases:
-
Chromosome Location:
Chr.15: 51982872 - 52029841 on Build Rnor_6.0
UniGene ID:
Rn.7037
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s221042

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc39a14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107275.1 NP_001100745.1 7 1047
XM_006252270.2 XP_006252332.1 7 1072

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

iron ion transport
zinc II ion transport
cellular zinc ion homeostasis
ferrous iron transport
iron ion transmembrane transport
zinc II ion transmembrane transport
zinc II ion transmembrane import
zinc ion transmembrane transporter activity
ferrous iron transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0