This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other REL SIRNA Assays ›
Gene Symbol:
Rel
Gene Name:
v-rel avian reticuloendotheliosis viral oncogene homolog
Gene Aliases:
-
Chromosome Location:
Chr.14: 108490794 - 108517589 on Build Rnor_6.0
UniGene ID:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s221035

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rel
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770448.1 XP_008768670.1 10 2009

Back To Top

More Information


Panther Classification:

Molecular Function -

Rel homology transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
cytokine production
transcription from RNA polymerase II promoter
inflammatory response
I-kappaB kinase/NF-kappaB signaling
response to cytokine
NIK/NF-kappaB signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of interleukin-12 biosynthetic process
innate immune response
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of neuron death
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0