This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WBP4 SIRNA Assays ›
Gene Symbol:
WBP4
Gene Name:
WW domain binding protein 4
Gene Aliases:
FBP21
Chromosome Location:
Chr.13: 41061561 - 41084006 on Build GRCh38
UniGene ID:
Hs.411300
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s22103

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: WBP4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007187.3 NP_009118.1 4 344
XM_005266245.2 XP_005266302.1 4 361
XM_017020380.1 XP_016875869.1 4 321
GenBank mRNA AF071185.1 - 4 380
AK000979.1 - 4 396
AK001557.1 - 4 379
AK297536.1 - 3 282
AK314525.1 - 4 381
BC104879.1 - 4 288 8143882
BC108310.1 - 4 345 6014541

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA cis splicing, via spliceosome
nucleic acid binding
protein binding
zinc ion binding
proline-rich region binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0