This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR1315 SIRNA Assays ›
Gene Symbol:
Olr1315 LOC103690273
Gene Name:
olfactory receptor 1315
olfactory receptor 149
Gene Aliases:
-
-
Chromosome Location:
Chr.8: 43469442 - 43470377 on Build Rnor_6.0
Chromosome Location:
Chr.8: 43523285 - 43524220 on Build Rnor_6.0
UniGene ID:
Rn.143482
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s220906

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr1315
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000469.1 NP_001000469.1 393
Gene Symbol: LOC103690273
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766127.1 XP_008764349.1 1 393

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm