This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE2A SIRNA Assays ›
Gene Symbol:
Ube2a
Gene Name:
ubiquitin-conjugating enzyme E2A
Gene Aliases:
-
Chromosome Location:
Chr.X: 123486994 - 123497726 on Build Rnor_6.0
UniGene ID:
Rn.8585
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s220836

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ube2a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013933.1 NP_001013955.1 5 343
XM_006257446.2 XP_006257508.1 4 356
GenBank mRNA BC079353.1 - 5 343 7108236

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019668, AK164074, BC026053, AK150417, AF089812, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
in utero embryonic development
DNA repair
biological_process
positive regulation of cell proliferation
response to UV
protein ubiquitination
histone ubiquitination
histone H2A ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein autoubiquitination
maternal process involved in female pregnancy
protein K48-linked ubiquitination
protein K11-linked ubiquitination
molecular_function
ubiquitin-protein transferase activity
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0