This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR668 SIRNA Assays ›
Gene Symbol:
Olr668 Olr665
Gene Name:
olfactory receptor 668
olfactory receptor 665
Gene Aliases:
-
-
Chromosome Location:
Chr.3: 77660928 - 77661863 on Build Rnor_6.0
Chromosome Location:
Chr.3: 77605027 - 77605962 on Build Rnor_6.0
UniGene ID:
Rn.142834
Rn.142836
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s220761

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr668
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000348.1 NP_001000348.1 1 755
Gene Symbol: Olr665
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000634.1 NP_001000634.1 1 755

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header