This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPSM1 SIRNA Assays ›
Gene Symbol:
Gpsm1
Gene Name:
G-protein signaling modulator 1
Gene Aliases:
Ags3
Chromosome Location:
Chr.3: 3767394 - 3794360 on Build Rnor_6.0
UniGene ID:
Rn.53933
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s220446

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpsm1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144745.2 NP_653346.2 6, 7 919
XM_006233670.2 XP_006233732.1 6, 7 1023
GenBank mRNA AF107723.1 - 5, 6 1043
CB615566.1 - 4, 5 475

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001199147, NM_001199146, NM_153410, AK082985, BC071197, and others.

Gene Ontology Categories:

Function(s) Process(es)

nervous system development
regulation of G-protein coupled receptor protein signaling pathway
cell differentiation
regulation of catalytic activity
G-protein alpha-subunit binding
GDP-dissociation inhibitor activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0