This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBXN11 SIRNA Assays ›
Gene Symbol:
Ubxn11
Gene Name:
UBX domain protein 11
Gene Aliases:
Soc, Ubxd5
Chromosome Location:
Chr.5: 152332993 - 152356647 on Build Rnor_6.0
UniGene ID:
Rn.11961
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s220416

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ubxn11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138853.2 NP_620208.1 12 1042
XM_006239000.1 XP_006239062.1 12 1104
XM_006239002.1 XP_006239064.1 10 978
XM_006239004.1 XP_006239066.1 10 958
XM_006239005.2 XP_006239067.1 9 1312
XM_008764131.1 XP_008762353.1 10 1110
XM_008764132.1 XP_008762354.1 10 1151
GenBank mRNA AB072920.1 - 11 970
BC078730.1 - 12 1042 7134156

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_183008, NM_145345, NM_001077262, AF521017, AK074358, and others.

Gene Ontology Categories:

Function(s) Process(es)

proteasome-mediated ubiquitin-dependent protein catabolic process
ubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0