This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GUCY2C SIRNA Assays ›
Gene Symbol:
Gucy2c
Gene Name:
guanylate cyclase 2C
Gene Aliases:
GCC
Chromosome Location:
Chr.4: 170659993 - 170740274 on Build Rnor_6.0
UniGene ID:
Rn.10931
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s220214

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gucy2c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013170.1 NP_037302.1 15, 16 1723
GenBank mRNA M55636.1 - 15, 16 1723

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
protein phosphorylation
response to toxic substance
intracellular signal transduction
regulation of cell proliferation
guanylate cyclase activity
protein kinase activity
ATP binding
GTP binding
toxic substance binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0