This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KPTN SIRNA Assays ›
Gene Symbol:
KPTN
Gene Name:
kaptin, actin binding protein
Gene Aliases:
2E4, MRT41
Chromosome Location:
Chr.19: 47475141 - 47486792 on Build GRCh38
UniGene ID:
Hs.25441
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s21956

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KPTN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291296.1 NP_001278225.1 4 524
NM_007059.3 NP_008990.2 6 692
XM_011526399.2 XP_011524701.1 9 2197
XM_017026226.1 XP_016881715.1 9 1042
XM_017026227.1 XP_016881716.1 8 1118
XM_017026228.1 XP_016881717.1 9 1042
GenBank mRNA AF105369.1 - 6 624
AK023996.1 - 6 647
AK225419.1 - 6 636
BC009249.2 - 6 607 3029623
DR759729.1 - 4 436

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

movement of cell or subcellular component
actin filament organization
sensory perception of sound
actin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0