This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB12 SIRNA Assays ›
Gene Symbol:
Rab12
Gene Name:
RAB12, member RAS oncogene family
Gene Aliases:
-
Chromosome Location:
Chr.9: 114683985 - 114709613 on Build Rnor_6.0
UniGene ID:
Rn.11021
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s217993

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013017.1 NP_037149.1 320
GenBank mRNA M83676.1 - 2 115

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_024448, AB232607, CK624286, M79303.

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle docking involved in exocytosis
autophagy
small GTPase mediated signal transduction
endosome to lysosome transport
protein secretion
protein transport
positive regulation of macroautophagy
regulation of exocytosis
Rab protein signal transduction
cellular response to insulin stimulus
cellular protein catabolic process
protein localization to plasma membrane
GTPase activity
GTP binding
GDP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header