This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ADM SIRNA Assays ›
Gene Symbol:
Adm
Gene Name:
adrenomedullin
Gene Aliases:
Ap, H39316, RATAP
Chromosome Location:
Chr.1: 175443208 - 175447260 on Build Rnor_6.0
UniGene ID:
Rn.10232
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s217919

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adm
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012715.1 NP_036847.1 4 437
XM_008759687.1 XP_008757909.1 5 1944
GenBank mRNA BC061775.1 - 4 433 5621783
D15069.1 - 4 419
U15419.1 - 4 437

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

vasculogenesis
response to hypoxia
neural tube closure
regulation of the force of heart contraction
G-protein coupled receptor internalization
cAMP biosynthetic process
positive regulation of cytosolic calcium ion concentration
heart development
female pregnancy
aging
androgen metabolic process
positive regulation of cell proliferation
negative regulation of cell proliferation
response to cold
response to wounding
response to organic substance
positive regulation of heart rate
cAMP-mediated signaling
positive regulation of cAMP biosynthetic process
organ regeneration
neuron projection regeneration
receptor internalization
response to lipopolysaccharide
response to insulin
odontogenesis of dentin-containing tooth
response to starvation
positive regulation of apoptotic process
negative regulation of vascular permeability
positive regulation of angiogenesis
negative regulation of vasoconstriction
positive regulation of vasodilation
hormone secretion
developmental growth
response to glucocorticoid
calcium ion homeostasis
branching involved in labyrinthine layer morphogenesis
spongiotrophoblast layer development
vascular smooth muscle cell development
positive regulation of vasculogenesis
hormone activity
protein binding
adrenomedullin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0