This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRNP SIRNA Assays ›
Gene Symbol:
Prnp
Gene Name:
prion protein
Gene Aliases:
PrP, Prn
Chromosome Location:
Chr.3: 124515922 - 124531320 on Build Rnor_6.0
UniGene ID:
Rn.3936
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s217881

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prnp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012631.2 NP_036763.1 3 798
XM_006235062.2 XP_006235124.1 4 950
GenBank mRNA BC072692.1 - 2 736 7099864
FQ211780.1 - 878
M20313.1 - 1 559

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001278256, NM_011170, M13685, AK157840, AK159224, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein phosphorylation
response to oxidative stress
learning or memory
negative regulation of interferon-gamma production
negative regulation of interleukin-17 production
negative regulation of interleukin-2 production
regulation of protein localization
cellular response to drug
negative regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of activated T cell proliferation
response to cadmium ion
response to copper ion
negative regulation of T cell receptor signaling pathway
protein homooligomerization
negative regulation of calcineurin-NFAT signaling cascade
cellular response to copper ion
regulation of potassium ion transmembrane transport
copper ion binding
microtubule binding
tubulin binding
identical protein binding
ATP-dependent protein binding
chaperone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0