This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRAFD1 SIRNA Assays ›
Gene Symbol:
TRAFD1
Gene Name:
TRAF-type zinc finger domain containing 1
Gene Aliases:
FLN29
Chromosome Location:
Chr.12: 112125545 - 112153604 on Build GRCh38
UniGene ID:
Hs.5148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s21435

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRAFD1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143906.1 NP_001137378.1 5 1215
NM_006700.2 NP_006691.1 5 684
GenBank mRNA AB007447.1 - 5 652
AK122620.1 - 5 1200
AK291331.1 - 5 669
AK309916.1 - 5 691
AK312601.1 - 5 684
BC003553.2 - 4 610 3547039

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to cytokine
negative regulation of innate immune response
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0