This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF460 SIRNA Assays ›
Gene Symbol:
ZNF460
Gene Name:
zinc finger protein 460
Gene Aliases:
HZF8, ZNF272
Chromosome Location:
Chr.19: 57279408 - 57298370 on Build GRCh38
UniGene ID:
Hs.99971
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s21206

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF460
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006635.3 NP_006626.3 3 727
XM_005258422.3 XP_005258479.1 3 1092
XM_017026176.1 XP_016881665.1 3 542
GenBank mRNA AK292144.1 - 3 727
AK298106.1 - 3 449
AY329492.1 - 3 648
BC109018.1 - 3 405
BC109019.1 - 3 405
BC118625.1 - 3 405 40006201

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header