This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RIIAD1 SIRNA Assays ›
Gene Symbol:
Riiad1
Gene Name:
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
Gene Aliases:
2310007A19Rik, Ncrna00166
Chromosome Location:
Chr.3: 94464984 - 94475078 on Build GRCm38
UniGene ID:
Mm.46405
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s211822

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Riiad1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025506.2 NP_079782.1 5, 6 400
GenBank mRNA AK009192.1 - 5, 6 402
AK019569.1 - 3, 4 210
BC107021.1 - 4, 5 787 40049310

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
calmodulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0