This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZC3H18 SIRNA Assays ›
Gene Symbol:
Zc3h18
Gene Name:
zinc finger CCCH-type containing 18
Gene Aliases:
1190001B23Rik, 5830416A07Rik, Nhn1
Chromosome Location:
Chr.8: 122376614 - 122417360 on Build GRCm38
UniGene ID:
Mm.241489
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s206703

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zc3h18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029993.1 NP_001025164.1 8 1380
NM_001029994.1 NP_001025165.1 7 1308
GenBank mRNA AK031413.1 - 8 1329
AK131189.1 - 5 452
AK159881.1 - 8 1316
AK165989.1 - 6 659
BC030495.1 - 7 1288 5400494
BC094330.1 - 5 461

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0