This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXO32 SIRNA Assays ›
Gene Symbol:
Fbxo32
Gene Name:
F-box protein 32
Gene Aliases:
4833442G10Rik, AI430017, ATROGIN1, Gm20361, MAFbx
Chromosome Location:
Chr.15: 58175879 - 58214892 on Build GRCm38
UniGene ID:
Mm.25683
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s205998

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxo32
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026346.3 NP_080622.1 6 761
GenBank mRNA AF441120.1 - 6 807
AK014612.1 - 6 714
AK014793.1 - 6 743
BC027211.1 - 6 695 3981597

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_133521, AY059628.

Gene Ontology Categories:

Function(s) Process(es)

response to electrical stimulus involved in regulation of muscle adaptation
muscle atrophy
response to denervation involved in regulation of muscle adaptation
cellular response to dexamethasone stimulus
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0