This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZCCHC18 SIRNA Assays ›
Gene Symbol:
Zcchc18
Gene Name:
zinc finger, CCHC domain containing 18
Gene Aliases:
1500031H04Rik, Sizn2
Chromosome Location:
Chr.X: 136992781 - 136996923 on Build GRCm38
UniGene ID:
Mm.440155
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s205699

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zcchc18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035509.1 NP_001030586.1 4 884
NM_001035510.1 NP_001030587.1 2 1355
NM_025893.2 NP_080169.2 4 832
GenBank mRNA AK005317.1 - 4 810
AK075607.1 - 1 436
AK134856.1 - 1213
AY650116.1 - 1 314
BC017627.1 - 4 745 4505033

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006257298.

Gene Ontology Categories:

Function(s) Process(es)

BMP signaling pathway
nucleic acid binding
zinc ion binding
ligand-dependent nuclear receptor transcription coactivator activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0