This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SCGB1B20 SIRNA Assays ›
Gene Symbol:
Scgb1b20 Scgb1b19 Scgb1b29 Scgb1b7
Gene Name:
secretoglobin, family 1B, member 20
secretoglobin, family 1B, member 19
secretoglobin, family 1B, member 29
secretoglobin, family 1B, member 7
Gene Aliases:
EG545948, Gm5895
Abpa19, EG434676, Gm5632
Abpa29, Abpa29_a7, EG384589, Gm5326
Abpa7, EG626305, Gm6662
Chromosome Location:
Chr.7: 33373234 - 33374559 on Build GRCm38
Chromosome Location:
Chr.7: 33287307 - 33288491 on Build GRCm38
Chromosome Location:
Chr.7: 32441541 - 32442844 on Build GRCm38
Chromosome Location:
Chr.7: 31712665 - 31713980 on Build GRCm38
UniGene ID:
Mm.349644
-
Mm.454021
Mm.454021
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
siRNA ID s203567

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Scgb1b20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270543.1 NP_001257472.1 2, 3 265
GenBank mRNA CK616096.1 - 2, 3 266
CK616412.1 - 2, 3 261
Gene Symbol: Scgb1b19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281526.1 NP_001268455.1 2, 3 249
Gene Symbol: Scgb1b29
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256066.1 NP_001242995.1 2, 3 249
GenBank mRNA GU269228.1 - 2, 3 249
Gene Symbol: Scgb1b7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270542.1 NP_001257471.1 2, 3 266
GenBank mRNA CK615999.1 - 2, 3 267

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header