This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RINL SIRNA Assays ›
Gene Symbol:
Rinl
Gene Name:
Ras and Rab interactor-like
Gene Aliases:
5830482F20Rik, 9930116N10Rik
Chromosome Location:
Chr.7: 28788861 - 28798965 on Build GRCm38
UniGene ID:
Mm.74632
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s203290

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rinl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177158.5 NP_796132.2 10 1447
GenBank mRNA AK030993.1 - 10 1440
AK082828.1 - 3 683
AK162469.1 - 3 352
AK168472.1 - 3 610
BC044761.1 - 10 1483
BC044878.1 - 1 164
BC046530.1 - 10 1483 5329601

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106241, XM_006228653, XM_006228654, XM_008759121, XM_006228657, and others.

Gene Ontology Categories:

Function(s) Process(es)

transport
endocytosis
protein transport
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header