This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SBSN SIRNA Assays ›
Gene Symbol:
Sbsn
Gene Name:
suprabasin
Gene Aliases:
1110005D19Rik, AI747508
Chromosome Location:
Chr.7: 30748971 - 30756134 on Build GRCm38
UniGene ID:
Mm.250717
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s203212

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sbsn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083903.1 NP_001077372.1 4 697
NM_172205.3 NP_757342.2 4 2251
GenBank mRNA AK003459.1 - 4 395
AY115494.1 - 4 2244
AY444556.1 - 4 636
BC029705.1 - 3 2433
BC051531.1 - 4 706
BC092350.1 - 4 626

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001198591, NM_001198590, NM_001044231, BC157808, BC107924, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0