This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLFR462 SIRNA Assays ›
Gene Symbol:
Olfr462 Olfr463
Gene Name:
olfactory receptor 462
olfactory receptor 463
Gene Aliases:
MOR240-3
MOR240-1
Chromosome Location:
Chr.11: 87888959 - 87889894 on Build GRCm38
Chromosome Location:
Chr.11: 87892987 - 87893922 on Build GRCm38
UniGene ID:
Mm.377484
Mm.377485
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s202929

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr462
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146411.2 NP_666523.2 1 204
GenBank mRNA BC108939.1 - 204 40041186
BC108940.1 - 205
Gene Symbol: Olfr463
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146413.2 NP_666525.2 1 204
GenBank mRNA BC106969.1 - 1 204 40041195
BC106970.1 - 1 202

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0