This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMPRSS11F SIRNA Assays ›
Gene Symbol:
Tmprss11f
Gene Name:
transmembrane protease, serine 11f
Gene Aliases:
4631408C20, 4732406D01Rik
Chromosome Location:
Chr.5: 86521226 - 86632424 on Build GRCm38
UniGene ID:
Mm.106867
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s202796

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmprss11f
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178730.3 NP_848845.1 9 1087
GenBank mRNA AK028447.1 - 9 1089
AK028587.1 - 9 1082
AK029032.1 - 9 1049
AK037173.1 - 8 931
BC132629.1 - 8 937 40130906

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008770104.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0