This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF214 SIRNA Assays ›
Gene Symbol:
Rnf214
Gene Name:
ring finger protein 214
Gene Aliases:
D130054N24Rik
Chromosome Location:
Chr.9: 45863532 - 45906906 on Build GRCm38
UniGene ID:
Mm.220945
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s202629

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf214
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178709.4 NP_848824.1 9 1748
GenBank mRNA AK028768.1 - 9 1383
AK030208.1 - 9 1380
AK032376.1 - 9 814
AK033867.1 - 9 1244
AK051527.1 - 9 1749
AK078706.1 - 4 282
AK136229.1 - 4 273
BC023073.1 - 1 26
BC049909.1 - 9 889
BC094247.1 - 8 1233 5717984

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001077239, NM_001278249, NM_207343, BC031347, AK300160, and others.
This siRNA also targets the following transcripts in Rat: XM_008766183, XM_008766186, XM_008766184, XM_008766185, XM_003750501, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0