This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NPM2 SIRNA Assays ›
Gene Symbol:
NPM2
Gene Name:
nucleophosmin/nucleoplasmin 2
Gene Aliases:
-
Chromosome Location:
Chr.8: 22023308 - 22036908 on Build GRCh38
UniGene ID:
Hs.131055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s20260

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NPM2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286680.1 NP_001273609.1 7 1133
NM_182795.1 NP_877724.1 6 719
XM_011544360.2 XP_011542662.1 5 1355
XM_011544362.2 XP_011542664.1 7 752
XM_011544363.2 XP_011542665.1 6 633
XM_011544364.2 XP_011542666.1 6 1042
XM_017012947.1 XP_016868436.1 4 1245
XM_017012948.1 XP_016868437.1 7 1352
XM_017012949.1 XP_016868438.1 6 1518
XM_017012950.1 XP_016868439.1 6 1226
GenBank mRNA AK094267.1 - 6 1494
AY262113.1 - 7 809
AY262114.1 - 6 719

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
regulation of exit from mitosis
single fertilization
embryo development
oocyte differentiation
covalent chromatin modification
positive regulation of catalytic activity
positive regulation of DNA replication
positive regulation of meiotic nuclear division
protein homooligomerization
chromatin binding
protein binding
enzyme binding
histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0