This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PEX16 SIRNA Assays ›
Gene Symbol:
Pex16
Gene Name:
peroxisomal biogenesis factor 16
Gene Aliases:
-
Chromosome Location:
Chr.2: 92374676 - 92381220 on Build GRCm38
UniGene ID:
Mm.202782
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s201777

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pex16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145122.2 NP_660104.2 8 1071
GenBank mRNA AK020743.1 - 8 781
AK143073.1 - 8 1073
AK162716.1 - 8 771
BC010822.1 - 8 812 4218547

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting to peroxisome
peroxisome organization
peroxisome membrane biogenesis
protein import into peroxisome matrix
protein to membrane docking
ER-dependent peroxisome organization
protein import into peroxisome membrane
protein localization to endoplasmic reticulum
protein C-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0