This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNA1 SIRNA Assays ›
Gene Symbol:
Kcna1
Gene Name:
potassium voltage-gated channel, shaker-related subfamily, member 1
Gene Aliases:
AI840627, Kca1-1, Kv1.1, MBK1, Mk-1, Shak, mceph
Chromosome Location:
Chr.6: 126636463 - 126645801 on Build GRCm38
UniGene ID:
Mm.40424
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s201555

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcna1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010595.3 NP_034725.3 2 2819
GenBank mRNA AK039559.1 - 2 1814
AK158065.1 - 2 1787
BC112970.2 - 2 1424 40048563
Y00305.1 - 1 2198

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_173095, X12589, M26161.

Gene Ontology Categories:

Function(s) Process(es)

startle response
transport
ion transport
potassium ion transport
regulation of muscle contraction
neuroblast proliferation
brain development
cell communication by electrical coupling
magnesium ion homeostasis
neuronal action potential
hippocampus development
neuronal signal transduction
cellular protein localization
regulation of ion transmembrane transport
regulation of membrane potential
neuromuscular process
detection of mechanical stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception of touch
protein homooligomerization
transmembrane transport
cellular response to magnesium ion
potassium ion transmembrane transport
ion channel activity
voltage-gated ion channel activity
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header